Learn More
Artefenomel, MedChemExpress
MedChemExpress Artefenomel (OZ439) is a synthetic antimalarial agent with the artemisinin pharmacophore. Artefenomel (OZ439) is a long-acting artemisinin-related agent.
Brand: Medchem Express HY-16762
This item is not returnable.
View return policy
Description
Artefenomel (OZ439) is a synthetic antimalarial agent with the artemisinin pharmacophore. Artefenomel (OZ439) is a long-acting artemisinin-related agent.Specifications
Artefenomel | |
1029939-86-3 | |
100 mg | |
C28H39NO5 | |
OZ439 | |
[H][C@@]1(C[C@H](C2)C3)C4([C@@]3([H])C[C@H]2C1)O[C@]5(OO4)CC[C@H](CC5)C6=CC=C(C=C6)OCCN7CCOCC7 | |
469.61 | |
Research |
Solid | |
Off-White | |
Research | |
COVID-19-immunoregulation | |
DMSO : 4.2 mg/mL (8.94 mM; Need ultrasonic) | |
469.61 | |
98.16% |
Product Suggestions
Customers who viewed this item also viewed
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.
Product Title
Artefenomel, MedChemExpress