Learn More
Artificial Seawater, Reagecon
BIOPHARMA
High-quality, pure water product, presented in a convenient pack size.
Brand:  Reagecon DSW5
 View more versions of this product
Product Code. 11533222
  • 1340.00 SEK / 5 L
Explore more special offers
Description
Description
  • Water Reference Standard, Water Reference Material
  • Can be used as a matrix for a variety of solutes or as a cleaning/sanitization solution
Specifications
Specifications
HDPE Container
5 L
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot NumberDW2524K1 Certificate TypeCertificate of Analysis Date19/12/2024 Catalog NumberDSW5
Lot NumberDW2523M1 Certificate TypeCertificate of Analysis Date11/01/2024 Catalog NumberDSW5
Lot NumberDW2523F1 Certificate TypeCertificate of Analysis Date13/09/2023 Catalog NumberDSW5
Lot NumberDW2523C1 Certificate TypeCertificate of Analysis Date04/04/2023 Catalog NumberDSW5
Lot NumberDW2523A1 Certificate TypeCertificate of Analysis Date19/01/2023 Catalog NumberDSW5
Product Title
Artificial Seawater, Reagecon > 5 L