Learn More
Tocris Bioscience™ AS 2444697
Potent and selective IRAK4 inhibitor
Brand: Tocris Bioscience™ 5430/50
This item is not returnable.
View return policy
Description
AS 2444697 is a potent and selective interleukin-1 receptor-associated kinase 4 (IRAK4) inhibitor (IC50 = 21 nM). Displays 30-fold selectivity for IRAK4 over IRAK1. Inhibits LPS-induced TNF-α and IL-6 production in PBMCs in vitro. Renoprotective and anti-inflammatory in a rodent model of chronic kidney disease.Specifications
N-[3-Aminocarbonyl)-1-(tetrahydro-2H-pyran-4-yl)-1H-pyrazol-4-yl]-2-(2-methyl-4-pyridinyl)-4-oxazolecarboxamide hydrochloride | |
1287665-60-4 | |
50 mg |