Learn More
ASTM (ASTM D6045, D1500) Colour Reference Standard, Paragon Scientific™
ASTM Colour Reference Standards are for routine calibration of colour measuring instruments and verification of test data. Process industries throughout the world use colorimeters for colour control of products such as edible and industrial oils, fuels, chemicals, pharmaceuticals, paints and coatings.
1860.00 SEK - 2150.00 SEK
Product Code | Brand | Description | Price | Quantity | Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Description | Price | Quantity | Availability | ||||
16040152
|
Paragon Scientific
134000 |
ASTM Color 1 | ASTM Color 1 | N/A | |||||
16050152
|
Paragon Scientific
134010 |
ASTM Color 3 | ASTM Color 3 | N/A | |||||
16060152
|
Paragon Scientific
134020 |
ASTM Color 5 | ASTM Color 5 | N/A | |||||
16070152
|
Paragon Scientific
134030 |
ASTM Color 7 | ASTM Color 7 | N/A | |||||
16050162
|
Paragon Scientific
134290 |
ASTM Color < 0.5 | ASTM Color < 0.5 | N/A | |||||
Description
- Certified according to ISO 17025 / ISO 17034 under UKAS accreditation
- Full Traceability to International Standards
- 500mL Pack Size supplied with MSDS
- 18 months shelf life
- Tamper evident security packaging
- Manufactured in the United kingdom
Specifications
500 mL | |
Color Reference Standard |
Safety and Handling
Product Identifier
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- ASTM Colour Reference Standard
Signal Word
- Danger
Hazard Category
- Aspiration toxicity Category 1
Hazard Statement
- H304-May be fatal if swallowed and enters airways.
Precautionary Statement
- P301+P310-IF SWALLOWED: Immediately call a POISON CENTER/doctor /
- P331-Do NOT induce vomiting.
- P405-Store locked up.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : white mineral oil
Nominal values quoted