Learn More
Astragaloside IV, MedChemExpress
MedChemExpress Astragaloside IV, an active component isolated from Astragalus membranaceus, suppresses the activation of ERK1/2 and JNK, and downregulates matrix metalloproteases (MMP)-2, (MMP)-9 in MDA-MB-231 breast cancer cells.
968.54 SEK - 3814.59 SEK
Specifications
Chemical Name or Material | Astragaloside IV |
---|---|
CAS | 84687-43-4 |
Color | White |
Purity Grade Notes | Research |
Molecular Formula | C41H68O14 |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
16478978
|
Medchem Express
HY-N0431 |
10 mg |
968.54 SEK
10mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16444837
|
Medchem Express
HY-N0431 |
50 mg |
2339.42 SEK
50mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
16479076
|
Medchem Express
HY-N0431 |
100 mg |
3814.59 SEK
100mg |
Please sign in to purchase this item. Need a web account? Register with us today! | |||||
Description
Astragaloside IV, an active component isolated from Astragalus membranaceus, suppresses the activation of ERK1/2 and JNK, and downregulates matrix metalloproteases (MMP)-2, (MMP)-9 in MDA-MB-231 breast cancer cells.Specifications
Astragaloside IV | |
White | |
C41H68O14 | |
O[C@H]1[C@H](O)[C@@H](O)[C@]([H])(O[C@@H]2C(C)(C)[C@@]([C@@H](O[C@]3([H])O[C@H](CO)[C@@H](O)[C@H](O)[C@H]3O)C[C@]4([H])[C@@]56CC[C@@]7(C)[C@@]4(C)C[C@H](O)[C@]7([H])[C@]8(C)O[C@H](C(C)(O)C)CC8)([H])[C@]5(C6)CC2)OC1 | |
784.97 | |
Research | |
Cancer-Kinase/protease |
84687-43-4 | |
Research | |
DMSO : ≥ 100 mg/mL (127.39 mM) | |
784.97 | |
98.0% | |
Solid |
Safety and Handling
missing translation for 'shelfLife' : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Recommended Storage : Powder -20°C 3 years, 4°C 2 years∣In solvent -80°C 6 months, -20°C 1 month
Research purposes only
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.