missing translation for 'onlineSavingsMsg'
Learn More
Learn More
ATP5L Rabbit anti-Human, Mouse, Rat, Polyclonal, Proteintech
Rabbit Polyclonal Antibody
Brand: Proteintech 16307-1-AP-150UL
This item is not returnable.
View return policy
Description
UCK2 catalyzes the phosphorylation of uridine monophosphate to uridine diphosphate. This is the first step in the production of the pyrimidine nucleoside triphosphates required for RNA and DNA synthesis. In addition, an allele of this gene may play a role in mediating nonhumoral immunity to Hemophilus influenzae type B.Specifications
ATP5L | |
Polyclonal | |
Unconjugated | |
ATP5L | |
ATP5JG, ATP5L, ATPase subunit g | |
Rabbit | |
Antigen Affinity Chromatography | |
RUO | |
10632, 27425, 300677 | |
-20°C | |
Liquid |
Immunohistochemistry (Paraffin), Western Blot | |
0.28 mg/mL | |
PBS with 50% glycerol and 0.02% sodium azide; pH 7.3 | |
O75964, Q6PDU7, Q9CPQ8 | |
Atp5l | |
ATP5L Fusion Protein Ag9287 | |
150 μL | |
Primary | |
Human, Mouse, Rat | |
Antibody | |
IgG |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction