Learn More
3M™ Attest™ Rapid Readout Biological Indicators
Monitors the effectiveness of the steam sterilization process and provides results within 3 hours. 3M™ Attest™ Rapid Readout Biological Indicators provide the best assurance of sterility by challenging the sterilizer with quantifiable, highly resistant spores.
Brand: 3M™ 7000053661
Additional Details : Weight : 0.63900kg
Description
- Easy to use and interpret with visual color readout
- Self-contained to significantly reduce the possibility of contamination
- Minimizes false positives
Specifications
Autoclavable | |
Brown | |
50 Pk. |
ISO 11138 | |
Biological Indicator | |
Steam |