Learn More
Invitrogen™ Attune™ NxT Flow Cell Cleaning Solution
The quality of your flow cytometry data can be affected by the cleanliness of the flow cell.
Brand:  Invitrogen™ A43635
Product Code. 16294971
  • 304.00 SEK / 30mL
Explore more special offers
Description
Description
Attune NxT Flow Cell Cleaning Solution is an alkaline liquid concentrate that, when mixed with water, removes contaminants without damaging the quartz glass used in the optical system of the Attune NxT Acoustic Focusing Cytometer.
Attune NxT Flow Cell Cleaning Solution has been formulated to minimize the contamination build-up that may occur in the system lines or flow cell after repeated usage. This solution is recommended for use during the SIP sanitize function just prior to the shutdown procedure. The recommended frequency of use depends upon the number of samples analyzed on the instrument. For high volume users, it is recommended as a daily preventive measure. For less frequent users, weekly use is recommended.
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Non-Interchangeable
Attune™ Acoustic Focusing Cytometer
30 mL
Cell Cleaning Solution
Store at room temperature
Attune™
Approved for shipment at Room Temperature or on Wet Ice
Certificates
A lot number is required to show results for certificates. To find your lot number on previous orders use our order status area.
5 results found
Lot Number Certificate Type Date Catalog Number
Lot Number3080117 Certificate TypeCertificate of Analysis Date17/12/2024 Catalog NumberA43635
Lot Number2994035 Certificate TypeCertificate of Analysis Date12/09/2024 Catalog NumberA43635
Lot Number2941327 Certificate TypeCertificate of Analysis Date12/06/2024 Catalog NumberA43635
Lot Number2878860 Certificate TypeCertificate of Analysis Date27/03/2024 Catalog NumberA43635
Lot Number2878859 Certificate TypeCertificate of Analysis Date14/03/2024 Catalog NumberA43635
Product Title
Invitrogen™ Attune™ NxT Flow Cell Cleaning Solution > Non-Interchangeable