missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Thermo Scientific Chemicals Auramine O, 80% dye content
Click to view available options
Quantity:
25 g
50 g
Description
Fluorescent stain for acid-resistant bacteria in sputum
This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.
Specifications
Specifications
Melting Point | 250°C (decomposition) |
UN Number | UN2811 |
Beilstein | 4030061 |
Solubility Information | Soluble in water or ethanol |
Formula Weight | 303.84 |
Color | Yellow |
Quantity | 50 g |
Physical Form | Powder |
Chemical Name or Material | Auramine O, 80% dye content |
RUO – Research Use Only
missing translation for 'provideContentCorrection'
missing translation for 'type03FormHeader'
missing translation for 'productTitle'