Learn More
Pacific Handy Cutter Auto-Retractable Metal Safety Knife
The QBAR is a particularly robust safety knife made of aluminum for the toughest applications! It has a fully automatic blade retraction and is therefore particularly safe.
Brand: Pacific Handy Cutter QBAR
This item is not returnable.
View return policy
Description
- In addition, it is equipped as standard with a blade with a rounded tip to reduce the risk of puncturing.
- The blade retracts fully automatically into the body, even if your thumb remains on the slider after the cut.
- Thanks to the three-sided ambidextrous slider, it is particularly ergonomic and can be used flexibly for horizontal and vertical cuts.
- The sturdy metal handle has non-slip rubber grips.
- Your knife is dirty? No problem. Thanks to the central lock, it can be opened and cleaned very easily without tools.
- Change the blade in the same way. V
- ery practically - you can accommodate replacement blades inside the handle.
- The QBAR can be attached or hung by means of an eyelet at the end of the handle.
- And the tape splitter on the underside of the knife head minimises use of your blade.
- Especially suitable for thicker cutting materials, corrugated cardboard, strapping, adhesive tape, foil, textiles, felt, PVC, leather, rubber, string, etc.
- QBAR is a premium quality safe cutting tool.
Specifications
Aluminum | |
Cutting Thicker Cutting Materials, Corrugated Cardboard, Strapping, Adhesive Tape, Foil, Textiles, Felt, PVC, Leather, Rubber, String, Etc. | |
Reusable | |
Non-sterile |