Learn More
Systec™ Autoclave
Use these benchtop and vertical autoclaves for a wide range of media sterilization applications, with options suitable for any budget. Systec™ Autoclaves are ideal for process optimization and validatable sterilization in laboratory applications.
Brand:  Systec™ 1523
 View more versions of this product
Product Code. 10542275
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Housing, support frame and pressure vessel made of corrosion-resistant stainless steel
  • Temperature and pressure range 140°C, 4 bar (except for Systec D-23)
  • LCD display and fully automatic microprocessor control
  • 3 to 25 sterilization programs depending on model
  • Flexible PT-100 temperature sensor
Systec V-Series Top-loading Vertical Autoclaves
  • Feature 8 different sizes with 40 to 150 liters chamber volume
  • Distinguished by its small footprint and high chamber capacity
  • Optimal loading capacity with most standard media bottles and Erlenmeyer flasks
Systec D-Series Front-loading Bench-top Autoclaves
  • Available in 7 sizes with chamber capacities ranging from 23 to 200 liters
  • Compact on the outside but with ideal chamber capacities inside
  • Each size has an optimal loading capacity for most standard media bottles and Erlenmeyer flasks
Specifications
Specifications
Autoclavable
25 L (Total), 23 L (Nominal)
260 mm
Autoclave, Front-loading Horizontal
230 V
RS-232/RS-485
3 to 10 bar
400 w/hr.
16 A
240 mm
DE-23
103 kg
50/60 Hz
80 kg
230 V