Learn More
Cole-Parmer™ Autoclave Deodorizer
Deoderize your autoclave with these capsules. Cole-Parmer™ Autoclave Deodorizers chemically absorb odor-causing molecules, removing unwanted odors from your facility rather than masking them.
Brand:  Cole-Parmer™ 44468-28
2522.25 SEK valid until 2025-04-30
Use promo code "24138" to get your promotional price.
Product Code. 11765868
  • 2655.00 SEK / Pack of 240
Explore more special offers
Description
Description
  • Lemon Scented Capsules
  • Absorb and remove autoclave odors rather than just covering them
  • Simply place one capsule in your autoclave or incubator with each load
  • The heat and moisture dissolves the capsule and releases a clean, fresh scent
Specifications
Specifications
Autoclave Deodorizers, Lemon; 40/Bottle, 6 Bottles/Cs
40/Bottle, 6 Bottles/Cs.
Special Offers
  • Multiple Manufacturers
    Use promotion code 24138 to get amazing savings on products for your analysis
    Promo Code: 24138
    Expires2025-04-30
    Days
    18
    Hours
    23
    Minutes
    34
    Seconds
    14
Product Content Correction
Product Title
Cole-Parmer™ Autoclave Deodorizer > Autoclave Deodorizer