Learn More
MP Biomedicals™ Avidin from Egg White
Avidin from Egg White, 10-15 u/mg solid, White Crystalline Powder
Brand:  MP Biomedicals™ 0215004780
Additional Details : CAS Number : 1405-69-2 Weight : 0.01000kg
 View more versions of this product
Product Code. 11486442
  • 12200.60 SEK / 100mg
Estimated Shipment: 06-08-2024
to see stock.
Add to basket
Description
Description
  • Avidin is useful in a biotin-avidin system for labeling biomolecules for receptor studies, immunoassays and immunohistological methods.
  • Avidin egg white was used in an assay using functionalized xenon as a biosensor to detect biotin-avidin binding.
  • Egg white was used at 80 nmol.
  • Egg white avidin has been used as a cross-linker for biotin-conjugated antibodies.
  • The product has also been used as a blocking agent in ELISA applications.
  • Avidin is a basic glycoprotein isolated from raw egg white.
  • It exhibits high binding affinity for biotin and is capable of producing biotin deficiency in rats and chicks.
  • It also occurs in the white portion of eggs and the oviducts of birds and amphibia.
  • It is destroyed by cooking or irradiation.
  • One unit will bind 1 ug of D-biotin at pH 8.9.
Specifications
Specifications
White
Egg White
-20°C
Labeling biomolecules for receptor studies, immunoassays and immunohistological methods
100 mg
1405-69-2
Crystalline