Learn More
Zeiss Microscope Carl Zeiss Axio Vert.A1 inverted for transmitted-light brightfield HAL and phase contra 560mm (w x d x h) 100-240V 50-60Hz 1.25x to 100xob
Brand: Zeiss 491237-0005-000
Microscope Carl Zeiss Axio Vert.A1 inverted for transmitted-light brightfield HAL and phase contra 560mm (w x d x h) 100-240V 50-60Hz 1.25x to 100xob
Additional Details : Weight : 10.50000kg