Learn More
Axygen™ Polypropylene Microtubes with Yellow Screw Cap
Brand:  Axygen SCT-200-Y
Product Code. 11314205
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Specifications
Screw Cap
2 mL
Screw Cap, O-ring
4000Pack
No
Yellow
Polypropylene
4000Pack
Non-sterile
Screw Cap
Documents
One moment while we fetch your results.
Product Title
Axygen™ Polypropylene Microtubes with Yellow Screw Cap > Capacity: 2mL; Quantity: 4000 Pack