Learn More
Axygen™ Polypropylene V-Bottom 384 Well Assay Plates
Brand: Axygen P-2ML-SQ-C-S-IND
Additional Details : Weight : 3.20000kg
Specifications
Standard | |
Clear | |
384 | |
Round Bottom (U-well) |
16 x 24 | |
Polypropylene | |
2 mL | |
Sample Collection |