missing translation for 'onlineSavingsMsg'
Learn More

STEMCELL Technologies AZ628​

MEK/ERK pathway inhibitor; Inhibits B-RAF, B-RAFV600E, and C-RAF-1

POA - 452.25 SEK

Specifications

CAS 878739-06-1
Molecular Formula C27H25N5O2
Purity ≥98%
View More Specs

Products 2
Product Code Brand Quantity Price Quantity & Availability  
Product Code Brand Quantity Price Quantity & Availability  
17160152
Encompass_Preferred
View Documents Promotion Details
STEMCELL Technologies
72982
1 mg
452.25 SEK
1mg
View alternative products
Save Up to 
»
Please to purchase this item. Need a web account? Register with us today!
17170152
Encompass_Preferred
View Documents Promotion Details
STEMCELL Technologies
72984
10 mg
POA
10mg
View alternative products
Save Up to 
»
N/A
Please to purchase this item. Need a web account? Register with us today!
This item requires a price update prior to being ordered.

Request a Price for this item here.

Description

Description

  • AZ628 is a quinazolinone that inhibits several Rapidly accelerated fibrosarcoma (RAF) kinases, including B-RAF, B-RAF V600E, and C-RAF-1 (in vitro kinase assay IC50 values of 105, 34, and 29 nM respectively; Khazak et al.) It strongly promotes B-Raf dimerization as a tight-binding inhibitor with a very slow off rate (Hatzivassiliou et al.; Lavoie et al.) From specificity profiling it is known to prevent activation of several other tyrosine kinases, including kinase insert domain receptor (KDR), discoidin domain receptor 2 (DDR2), Lck/Yes Novel (LYN), feline McDonough sarcoma (FMS), and FMS-like tyrosine kinase 1 (FLT1) (Khazak et al.)
  • Inhibits tumor growth, induces cell cycle arrest and causes apoptosis in a variety of cancer cell lines, especially in lines with the B-RAF V600E mutation (McDermott et al.; Khazak et al.)
Specifications

Specifications

878739-06-1
C27H25N5O2
≥98%
Videos
SDS
Documents

Documents

Product Certifications