Learn More
AZD7507, MedChemExpress
MedChemExpress AZD7507 is a potent and orally active CSF-1R inhibitor, with antitumor activity.
Brand: Medchem Express HY-117244
This item is not returnable.
View return policy
Description
AZD7507 is a potent and orally active CSF-1R inhibitor, with antitumor activity.Specifications
AZD7507 | |
Yellow | |
C23H27FN6O3 | |
DMSO : 130 mg/mL (286.03 mM; Need ultrasonic) | |
454.5 | |
99.14% | |
Solid |
1041852-85-0 | |
Research | |
10 mg | |
O=C(N)C1=NN=C2C=C(C(N3CCN(CC3)CCO)=CC2=C1NC4=CC=C(C=C4F)C)OC | |
454.5 | |
Research | |
Cancer-Kinase/protease |
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.