Learn More
AZD7507, MedChemExpress
MedChemExpress AZD7507 is a potent and orally active CSF-1R inhibitor, with antitumor activity.
Brand:  Medchem Express HY-117244
Encompass_Preferred
 View more versions of this product
Product Code. 16484868
  • 4823.79 SEK / 10mg
Explore more special offers
Description
Description
AZD7507 is a potent and orally active CSF-1R inhibitor, with antitumor activity.
Specifications
Specifications
AZD7507
Yellow
C23H27FN6O3
DMSO : 130 mg/mL (286.03 mM; Need ultrasonic)
454.5
99.14%
Solid
1041852-85-0
Research
10 mg
O=C(N)C1=NN=C2C=C(C(N3CCN(CC3)CCO)=CC2=C1NC4=CC=C(C=C4F)C)OC
454.5
Research
Cancer-Kinase/protease
Product Content Correction
The Fisher Scientific Encompass Program offers items which are not part of our distribution portfolio. These products typically do not have pictures or detailed descriptions. However, we are committed to improving your shopping experience. Please use the form below to provide feedback related to the content on this product.