missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Azelaic Acid, MP Biomedicals
Click to view available options
100g
Specifications
CAS | 123-99-9 |
Molecular Formula | C9H16O4 |
Synonym | azelaic acid, finacea, anchoic acid, azelex, lepargylic acid, skinoren, 1,7-heptanedicarboxylic acid, heptanedicarboxylic acid, 1,9-nonanedioic acid, emerox 1110 |
InChI Key | BDJRBEYXGGNYIS-UHFFFAOYSA-N |
SMILES | C(CCCC(=O)O)CCCC(=O)O |
IUPAC Name | nonanedioic acid |
Molecular Weight (g/mol) | 188.223 |
PubChem CID | 2266 |
ChEBI | CHEBI:48131 |
Formula Weight | 188.22 |
Show More |
missing translation for 'provideContentCorrection'
missing translation for 'productTitle'