Learn More
Azlon™ Gas Delivery Tube
Polypropylene construction. Azlon™ Gas Delivery Tube are ideal substitutes for fragile glass alternatives.
Brand: Azlon DT004
This item is not returnable.
View return policy
Description
- Pre-bent rigid polypropylene delivery tubes
- Unbreakable in normal use
- Suitable for many types of gas collection experiments in schools and universities
Specifications
Straight Tube | |
500 mm | |
-20°C | |
135°C |
Polypropylene | |
Many types of gas collection experiments in schools and universities | |
5 |