Learn More
Azlon™ Gas Delivery Tube
Polypropylene construction. Azlon™ Gas Delivery Tube are ideal substitutes for fragile glass alternatives.
Brand:  Azlon DT004
Product Code. 12483601
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products »
Explore more special offers
Description
Description
  • Pre-bent rigid polypropylene delivery tubes
  • Unbreakable in normal use
  • Suitable for many types of gas collection experiments in schools and universities
TRUSTED_SUSTAINABILITY
Specifications
Specifications
Straight Tube
500 mm
-20°C
135°C
Polypropylene
Many types of gas collection experiments in schools and universities
5