Learn More
Azlon™ HDPE Pipette Rack
White moulded polyethylene rack that will support 12 pipettes over 200mm long
Brand: Azlon SWP038
This item is not returnable.
View return policy
Description
- White molded polyethylene rack
- Will support 12 pipettes over 200mm long
- Very stable
- Supplied as a kit of two end pieces and three connecting rods for easy push fit assembly
Specifications
Pipette Rack | |
White | |
Pipettes (12 over 200 mm long) | |
HDPE |