Learn More
Azlon™ Rubber Pipette Fillers
Suitable for pipettes up to 100mL
275.00 SEK - 308.00 SEK
Product Code | Brand | Grip Type | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Grip Type | Price | Quantity & Availability | |||||
10507055
|
Azlon
PWY144 |
For all sizes |
275.00 SEK
Each |
In Stock
Log in to see stock. |
|||||
10430295
|
Azlon
PWY145 |
Single-handed |
293.00 SEK
Each |
Estimated Shipment: 21-08-2024
Log in to see stock. |
|||||
15718047
|
Azlon
PWY146 |
Extended insert nozzle for improved grip |
308.00 SEK
Each |
Estimated Shipment: 12-09-2024
Log in to see stock. |
|||||
Description
Suitable for pipettes up to 100mL
- Suitable for pipettes up to 100mL
- Pipette filler works via a series of glass ball valves for air release, suction and delivery
- PWY145 is suitable for single handed operation
- PWY146 has an extended insert nozzle for improved grip
Specifications
Rubber | |
Pipette Filler |