Learn More
Brady™ B33 Series Coated Vinyl Cloth Labels with Repositionable Adhesive
Permanent or repositionable, the B33 Series Vinyl Cloth Label is there to stay...unless you want to change it.
1805.00 SEK
Specifications
Adhesive Type | Repositionable Rubber |
---|---|
Color | White |
Finish | Semi-gloss |
Format | Die-cut |
Material | Vinyl Cloth |
Product Code | Brand | Height (English) | Labels per Row | Width (English) | For Use With (Application) | Price | Quantity & Availability | ||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Height (English) | Labels per Row | Width (English) | For Use With (Application) | Price | Quantity & Availability | ||
30041430
|
Brady™
153080 |
0.75 in. | 1 Label | 0.75 in. | Barcode Labeling, Equipment Labeling, Laboratory Labeling, Wire and Cable Labeling |
1805.00 SEK
Pack of 1000 |
Please sign in to purchase this item. Need a web account? Register with us today! | ||
Description
Permanent or repositionable, the B33 Series Vinyl Cloth Label is there to stay...unless you want to change it. The adhesive and cloth backing offers excellent holding power, while also allowing for clean removal and repositioning.
A ribbon is required to print this material.
- Sticks to any clean and dry surface
- Specially formulated topcoat for excellent print quality
- Semi-gloss finish for a crisp, clean look
Specifications
Repositionable Rubber | |
Semi-gloss | |
Vinyl Cloth | |
Thermal Transfer | |
Brady™ Label Printers | |
0.0073 in. |
White | |
Die-cut | |
Single-side Printable | |
Non-sterile | |
-40°F to 176°F |