Learn More
Invitrogen™ Bandmate™ Reagent Tubes
Bandmate Reagent Tubes are specially designed for use with the Bandmate Automated Western Blot Processor.
1584.00 SEK - 6410.00 SEK
Specifications
Color | Clear |
---|---|
For Use With (Application) | Western Blot |
For Use With (Equipment) | Invitrogen Bandmate Automated Western Blot Processor |
Product Line | Bandmate |
Type | Reagent Tube |
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Price | Quantity & Availability | |||||
17193309
|
Invitrogen™
BW020X25 |
500 tubes |
6410.00 SEK
Pack of 500 |
In Stock
Log in to see stock. |
|||||
17103319
|
Invitrogen™
BW020X5 |
100 tubes |
1584.00 SEK
Pack of 100 |
In Stock
Log in to see stock. |
|||||
Description
The tubes are required for dispensing primary and secondary antibodies into the Bandmate mini and midi incubation trays during the automated western blot processing procedure. Two reagent tubes are required for each blot.Specifications
Clear | |
Invitrogen Bandmate Automated Western Blot Processor | |
Reagent Tube |
Western Blot | |
Bandmate | |
Ambient |