Learn More
Thermo Scientific™ Barnstead™ GenPure™ UV/UF with installation
Produce Type 1 ultrapure water with consistent quality for your most demanding and sensitive applications with the Thermo Scientific™ Barnstead™ GenPure™ system.
Brand:  Thermo Scientific™ 50131217INS
Additional Details : Weight : 0.80000kg
Product Code. 16480831
  • 105912.15 SEK / Each
Estimated Shipment: 11-10-2024
to see stock.
Add to basket
Description
Description
Integrated feed water monitoring:
  • An additional measuring cell monitors feed water conductivity.
  • Any exceeding of the limiting value is immediately displayed maximizing cartridge life.
  • Online TOC measurement with uV intensity monitoring:
  • Real-time TOC monitoring allows continual measurement of organic substances present in the water.
  • Continuous monitoring of UV intensity removes the risk of false TOC values due to diminished UV intensity.
  • Fault message alerts user when the UV lamp intensity is no longer sufficient for accurate TOC measurements.
  • System offers UV Photo-oxidation 185/254nm.
  • High performance UV assembly reliably reduces the content of microorganisms and their metabolites.
  • Ready-to-use:
  • Feed water pressure switch, wall bracket, UV lamp, ultrapure cartridge, sterile filter, all included with one part number.
  • Measurement of conductivity/resistivity
  • Conductivity cells are carefully calibrated prior to each measurement via built-in reference resistance with cell constants at 0.01cm-1.
  • Temperature measurements are made by a platinum chip sensor with ± 0.1°C accuracy
  • Quick-connect cartridge replacement
  • Aquastop quick-connect capability enables cartridge replacement within seconds even mid-operation
  • GLP-compliant documentation
  • Real-time clock and code-protected operating system prevents unauthorized changes to system settings
  • RS-232 interface with adjustable send—interval for safe data transfer of all measured data, faults, date and time to a PC computer or log printer
  • Digital microprocessor control automatically monitors and stores faults from the last four weeks
  • USP-compliant conductivity measurement with temperature compensation can be switched on or off
  • Tilting control panels for easy viewing
  • The ergonomically designed controller tilts for optimal key pressing and easy readability
  • Illuminated four-line alpha-numeric display
  • Specifications
    Specifications
    0.055μS/cm
    <0.1 CFU/mL
    R 3/4 in.
    Pretreated
    18.2 Mohm.cm
    1 to 5 ppb
    2 to 6 bar
    0.1 kW
    372 mm
    330 mm
    90/240 V 50/60 Hz
    50/60 Hz
    GenPure ultrapure water system with UV-photo-oxidation and ultrafiltration membrane
    0.001 EU/mL
    Yes
    Up to 2 L/min.
    No
    1 to 6 bar
    <1
    2°C to 35°C
    615 mm
    23 kg
    90/240 V
    Special Offers
    Services