missing translation for 'onlineSavingsMsg'
Learn More

Base-Neutrals Matrix Spike Tinted Indicator, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

Brand:  SPEX Certiprep CLPS-MSB-TI

Additional Details : CAS Number : 67-56-1 Weight : 0.01000kg Transport : UN number : 1230 Chem class : 3 Pack group : II


Product Code. 15215976

  • 540.00 SEK / 1mL
Estimated Shipment: 03-09-2024
to see stock.

Includes: Autosampler Vial, NIST Traceable Certificate of Analysis accredited to ISO/IEC 17025 and ISO/IEC Guide 34, Safety Data Sheet

Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2,4-Trichlorobenzene120-82-11000μg/mL
1,4-Dichlorobenzene106-46-71000μg/mL
2,4-Dinitrotoluene121-14-21000μg/mL
Acenaphthene83-32-91000μg/mL
Bromophenol blue115-39-91000μg/mL
N-Nitrosodi-n-propylamine621-64-71000μg/mL
Pyrene129-00-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Base-Neutrals Matrix Spike Tinted Indicator
2mL amber prescored glass ampule
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
1000μg/mL
Methanol
120-82-1,106-46-7,121-14-2,83-32-9,115-39-9,621-64-7
1 mL
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
1,2,4-Trichlorobenzene
Product Suggestions

Product Suggestions

Videos
Safety and Handling

Safety and Handling

Product Identifier
  • BASE-NEUTRALS MATRIX SPIKE

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 3
  • Carcinogenicity Category 1B
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity after single exposure Category 1

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H331-Toxic if inhaled.
  • H350-May cause cancer.
  • H361-Suspected of damaging fertility or the unborn child.
  • H370-Causes damage to organs.

Precautionary Statement
  • P101-If medical advice is needed, have product container or label at hand.
  • P102-Keep out of reach of children.
  • P103-Read label before use.
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P260-Do not breathe dust/fume/gas/mist/vapours/spray.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : nitrosodipropylamine, methanol

SDS
Documents

Documents

Certificates
Special Offers

Special Offers

Product intended for laboratory use only