missing translation for 'onlineSavingsMsg'
Learn More

Base/Neutrals Surrogate Standard, SPEX CertiPrep™

ISO/IEC Guide 34 Certified, NIST-Traceable Certified Reference Material

930.00 SEK - 1350.00 SEK

Specifications

Chemical Name or Material Base/Neutrals Surrogate Standard
CAS 2199-69-1, 321-60-8, 4165-60-0, 1718-51-0
Concentration or Composition Notes Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
For Use With (Equipment) GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
Concentration or Composition (by Analyte or Components) 1000μg/mL
View More Specs

Products 2
Product Code Brand Quantity Packaging Price Quantity & Availability  
Product Code Brand Quantity Packaging Price Quantity & Availability  
12769695
View Documents Promotion Details
SPEX Certiprep
CLP90-SB
1 mL 2mL amber prescored glass ampule This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
N/A
12779695
View Documents Promotion Details
SPEX Certiprep
CLP90-SB5
5 mL 5mL amber prescored glass ampule
930.00 SEK
5mL
Please to purchase this item. Need a web account? Register with us today!
Description

Description

Organic Standard manufactured for GC, GC/MS, HPLC, LC/MS, and other analytical instrumentation

AnalyteCAS No.Concentration
1,2-Dichlorobenzene-d42199-69-11000μg/mL
2-Fluorobiphenyl321-60-81000μg/mL
Nitrobenzene-d54165-60-01000μg/mL
p-Terphenyl-d141718-51-01000μg/mL

Certifications

Accredited by A2LA for ISO/IEC 17025 and ISO/IEC Guide 34 and certified by UL-DQS to ISO 9001

Compliance

GHS/OSHA Complaint

Specifications

Specifications

Base/Neutrals Surrogate Standard
Certificate of Analysis reports target analytes to a +/−2% accuracy and correct for raw material purity.
1000μg/mL
MeCl:Acetone
2199-69-1, 321-60-8, 4165-60-0, 1718-51-0
GC, GC/MS, HPLC, LC/MS, and Other Analytical Instrumentation
1,2-Dichlorobenzene-d4,2-Fluorobiphenyl
Videos
Safety and Handling

Safety and Handling

Highly flammable liquid and vapor. Causes serious eye irritation. May cause drowsiness or dizziness.

missing translation for 'dotInformation' : Nonhazardous

Recommended Storage : Ambient

Signal Word
  • Danger

Hazard Category
  • Acute toxicity Category 4
  • Carcinogenicity Category 2
  • Serious eye damage/eye irritation Category 2
  • Flammable liquid Category 2
  • Reproductive toxicity Category 2
  • Specific target organ toxicity Category 3

Hazard Statement
  • H225-Highly flammable liquid and vapour.
  • H302-Harmful if swallowed.
  • H319-Causes serious eye irritation.
  • H336-May cause drowsiness or dizziness.
  • H351-Suspected of causing cancer.
  • H361-Suspected of damaging fertility or the unborn child.

Precautionary Statement
  • P210-Keep away from heat, hot surfaces, sparks, open flames and other ignition sources. No smoking.
  • P241-Use explosion-proof electrical/ventilating/lighting/ equipment.
  • P303+P361+P353-IF ON SKIN (or hair): Take off immediately all contaminated clothing. Rinse skin with water/ shower.
  • P305+P351+P338-IF IN EYES: Rinse cautiously with water for several minutes. Remove contact lenses, if present and easy to do. Continue rinsing.
  • P405-Store locked up.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : dichloromethane, acetone, nitrobenzene

SDS
Documents

Documents

Product Certifications

Product intended for laboratory use only