Learn More
Scharlab BAT Agar
BAT Agar is used for the detection and isolation of Alicyclobacillus in fruit juices and other acidic food.
Brand: Scharlab 01-675-500
This item is not returnable.
View return policy
Description
- IFU
- Solid medium for the detection and isolation of Alicyclobacillus, in fruit juices and other acidic food, according to IFU Standard Method No. 12.
- For detection of the species A. acidoterrestris recommend using the K Agar medium (Ref.: 01-674).
Specifications
Solid | |
Agar | |
Detection and Isolation of Alicyclobacillus | |
3 to 5 Days | |
500 g |
BAT Agar | |
Yeast Extract 2.00 g/L, Dextrose 5.00 g/L, Potassium Phosphate 3.00 g/L, Calcium Chloride 0.25 g/L, Magnesium Sulfate 0.50 g/L, Ammonium Sulfate 0.20 g/L, Zinc Sulfate 0.0002 g/L, Copper Sulfate 0.0002 g/L, Manganese Sulfate 0.0001 g/L, Sodium Molybdate 0.0003 g/L, Agar 20.00 g/L | |
45°C ± 1.0°C | |
4.2 ±0.2 |