missing translation for 'onlineSavingsMsg'
Learn More
Learn More
Brannan™ Batch Sampling Freezer Thermometer with Calibration Certificate
Precision test batch sampling freezer thermometers sealed into a 30 mL polypropylene bottle containing a heat transfer medium, ideally suited to monitor the temperature of samples in a controlled storage. Complete with traceable calibration.
1920.00 SEK - 2425.00 SEK
Specifications
Bottle Fill | Customer to fill with monopropylene glycol or other heat transfer medium before use |
---|---|
For Use With (Application) | Monitor temperature of samples in a controlled storage |
Height (English) | 5.71 in. |
Height (Metric) | 14.5 cm |
Includes | Polypropylene bottle and glass thermometer |
Product Code | Brand | Calibrated | Price | Quantity & Availability | ||||||
---|---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Calibrated | Price | Quantity & Availability | ||||||
17823058
|
Brannan™
22/101/2 + 49/101/0 |
3 Point Traceable |
1920.00 SEK
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
17863058
|
Brannan™
22/101/2 + 49/503/0 |
3-point UKAS |
2425.00 SEK
Each |
Estimated Shipment: 09-05-2025
Log in to see stock. |
||||||
Description
- Non-hazardous LO-tox™ filling
- Bottle should be filled with monpropylene glycol or other heat transfer medium before use
- Temperature measurement unaffected by short term fluctuations
- Supplied with calibration
- Temperature range: -30°C to 0°C
- Supplied with statement of accuracy
- Made in UK
Specifications
Customer to fill with monopropylene glycol or other heat transfer medium before use | |
5.71 in. | |
Polypropylene bottle and glass thermometer | |
Glass | |
1.9577 oz. | |
0.14 in. | |
0°C | |
-30°C | |
-30°C to 0°C | |
None | |
0.5°C | |
3.5 cm |
Monitor temperature of samples in a controlled storage | |
14.5 cm | |
14.5 cm | |
No | |
55.5 g | |
32°F | |
-22°F | |
-22°F to +32°F | |
Freezer | |
± 0.7°C | |
None |
Spot an opportunity for improvement?Share a Content Correction
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Brannan™ Batch Sampling Freezer Thermometer with Calibration Certificate