Learn More
BD 2001 Luminometer Cuvettes
For luminometer
Brand: BD 550768
Additional Details : Weight : 1.54570kg
Description
- Dimension: 12 x 47mm
Specifications
Luminometer Cuvette | |
Plastic | |
Each | |
For luminometer |
Research use only.