Learn More
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Rhamnose
L-rhamnose
Brand: BD 217520
This item is not returnable.
View return policy
Description
Specifications
ISO/DIS, 11014 | |
Rhamnose | |
Industrial microbial testing | |
100 g |
<30°C | |
Powder | |
Bottle |
Product Content Correction
Product Title
BD BBL™/Difco™ Media Additives/Ingredients/Reagents: Rhamnose > Rhamnose; BD Difco; 100g
Spot an opportunity for improvement?Share a Content Correction