Learn More
BD BBL™/Difco™ TB Decolorizer TM
For staining mycobacteria by the Truant, Brett and Thomas and the Morse, Blair, Weiser and Sproat fluorescent procedures
Brand:  BD 212512
Additional Details : CAS Number : 67-63-0 Weight : 0.27000kg
Product Code. 17342461
  • 2160.00 SEK / Pack of 4
Estimated Shipment: 01-10-2024
to see stock.
Add to basket
Description
Description
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
Specifications
Specifications
Decolorizer
For staining mycobacteria by the Truant, Brett and Thomas and the Morse, Blair, Weiser and Sproat fluorescent procedures
Bottle
4 x 250mL
15° to 30°C
Decolorizer TM
Product Suggestions
Product Suggestions
Videos
SDS
Documents
Documents
Certificates
Promotions
Promotions
For in vitro Diagnostic Use.