Learn More
BD BBL™/Difco™ TB Decolorizer TM
For staining mycobacteria by the Truant, Brett and Thomas and the Morse, Blair, Weiser and Sproat fluorescent procedures
Brand: BD 212512
Additional Details : CAS Number : 67-63-0 Weight : 0.27000kg
Description
Stains smears prepared from specimens and cultures suspected of containing mycobacteria for early presumptive diagnosis of mycobacterial infection and characterization of bacterial isolates.
Specifications
Decolorizer | |
For staining mycobacteria by the Truant, Brett and Thomas and the Morse, Blair, Weiser and Sproat fluorescent procedures | |
Bottle |
4 x 250mL | |
15° to 30°C | |
Decolorizer TM |
For in vitro Diagnostic Use.