missing translation for 'onlineSavingsMsg'
Learn More

BD BBL™ Mycoflask™ Prepared Tubed Media: Bile Esculin Agar Slants

Used to differentiate enterococci and the Streptococcus bovis group from other streptococci

4605.00 SEK

Specifications

Certifications/Compliance CE, COMPF
Color Beige
Content And Storage 2°C to 8°C
Form Solid
Manufacturing Origin United States
View More Specs

Products 1
Product Code Brand Quantity For Use With (Application) Price Quantity & Availability  
Product Code Brand Quantity For Use With (Application) Price Quantity & Availability  
17302331
View Documents
BD
221410
100/Pk. Isolation and cultivation of pathogenic and nonpathogenic fungi
4605.00 SEK
Pack of 100
Estimated Shipment: 03-02-2025
to see stock.
Add to basket
 
Description

Description

  • For cultures requiring prolonged incubation

Specifications

Specifications

CE, COMPF
2°C to 8°C
United States
Pancreatic digest of Gelatin 5g/L, Beef extract 3g/L, Oxgall 20g/L, Ferric Citrate 0.5g/L, Esculin 1g/L, Agar 14g/L
Beige
Solid
Bile Esculin Agar
Tube
Videos
Safety and Handling

Safety and Handling

missing translation for 'shelfLife' : 365 Days

Product Identifier
  • BD BBL Bile Esculin Agar Slants , 100 pack 6.8 Ml

Signal Word
  • Warning

Hazard Category
  • Skin sensitiser Category 1

Hazard Statement
  • H317-May cause an allergic skin reaction.

Precautionary Statement
  • P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
  • P272-Contaminated work clothing should not be allowed out of the workplace.
  • P280-Wear protective gloves/protective clothing/eye protection/face protection.
  • P302+P352-IF ON SKIN: Wash with plenty of water/.
  • P321-Specific treatment (see on this label).
  • P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
  • P362+P364-Take off contaminated clothing and wash it before reuse.
  • P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.

Supplemental information
  • MIXTURE LIST-Contains : bile extract

SDS
Documents

Documents

Product Certifications