Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Minocyline
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
Brand:  BD 231251
Additional Details : Weight : 0.80000kg
 View more versions of this product
Product Code. 11783819
  • 600.00 SEK / Pack of 500
Estimated Shipment: 13-09-2024
to see stock.
Add to basket
Description
Description
  • Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Specifications
Minocycline
30μg
Disc
10/Pk.