Learn More
BD BBL™ Sensi-Disc™ Susceptibility Test Discs: Moxifloxacin83
High-quality absorbent paper discs impregnated with accurately determined amounts of antibiotic or other chemotherapeutic agents
Brand: BD 231758
Additional Details : Weight : 0.01040kg
Description
- Discs containing certifiable agents are submitted to FDA for assay and certification before distribution Clearly marked on both sides with letters and numbers designating agent and concentration
Specifications
Moxifloxacin83 | |
5μg | |
Disc | |
10 Pack |