missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD BD Biosciences™ Polyethylene Perfusion Luer Lock Syringe
Brand: BD 300137
Additional Details : Weight : 0.20000kg
Specifications
Polyethylene | |
Perfusion Luer Lock Syringe | |
Sterile |
121 mm | |
50 mL |