Learn More
BD Cytometric Bead Array (CBA) Human CD54 (ICAM-1); Flex Set, Bead A4
Bead-based immunoassay capable of measuring soluble human soluble CD54, also known as ICAM-1, in serum, plasma, and cell culture supernatant samples
Brand:  BD 560269
Additional Details : Weight : 0.04000kg
Product Code. 15810069
  • 5965.00 SEK / Each
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Includes:
  • Human sCD54 (ICAM-1), Standard
  • Human sCD54 (ICAM-1), PE Detection Reagent
  • Human sCD54 (ICAM-1), Capture Bead A4
Description
Description
Contains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. The Standard component is lyophilized and should be transferred to a 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL Assay Diluent, the standard has a protein concentration of 10,000pg/mL.
  • Human reactivity was determined by testing samples with CBA sCD54 (ICAM-1) Flex Set
  • Must be used in conjunction with CBA Human Soluble Protein Master Buffer Kit, flow cytometer, and FCAP Array Software
Specifications
Specifications
CD54 (ICAM-1)
Human sCD54 (ICAM-1), standard, Human sCD54 (ICAM-1), PE detection reagent, Human sCD54 (ICAM-1), capture bead A4
4°C
Human CD54 (ICAM-1) flex set (bead A4)
Human
100 Tests
Flow Cytometry
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning