Learn More
BD Cytometric Bead Array (CBA) Human IL-13; Flex Set, Bead E6
Bead-based immunoassay for measuring human interleukin-13 (IL-13) in serum, plasma and cell culture supernatant samples
Brand:  BD 558450
Product Code. 15839398
  • 4630.00 SEK / Each
Estimated Shipment: 04-02-2025
to see stock.
Explore more special offers
Add to basket
Includes: Human IL-13 Standard (two vials), Human IL-13 PE Detection Reagent (one vial), Human IL-13 Capture Bead E6 (one vial)
Description
Description
  • Human reactivity was determined by testing samples with Flex Set
  • Theoretical limit of detection is 0.6pg/mL
  • Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components
Flow Cytometry
TRUSTED_SUSTAINABILITY
Specifications
Specifications
IL-13
Human IL-13 standard (2 vials), Human IL-13 PE detection reagent (one vial), Human IL-13 capture bead E6 (one vial)
Serum, Plasma, Cell Culture Supernatant
Flow Cytometry
Human
100 Tests
4°C
Human IL-13 flex set (bead E6)