Learn More
BD Cytometric Bead Array (CBA) Human IL-13; Flex Set, Bead E6
Bead-based immunoassay for measuring human interleukin-13 (IL-13) in serum, plasma and cell culture supernatant samples
Brand: BD 558450
Additional Details : Weight : 0.03600kg
Includes: Human IL-13 Standard (two vials), Human IL-13 PE Detection Reagent (one vial), Human IL-13 Capture Bead E6 (one vial)
Description
- Human reactivity was determined by testing samples with Flex Set
- Theoretical limit of detection is 0.6pg/mL
- Storage buffer: Aqueous buffered solution containing BSA/fetal bovine serum along with other reagents depending on kit components
Flow Cytometry
Specifications
IL-13 | |
Human IL-13 standard (2 vials), Human IL-13 PE detection reagent (one vial), Human IL-13 capture bead E6 (one vial) | |
Serum, Plasma, Cell Culture Supernatant | |
Flow Cytometry |
Human | |
100 Tests | |
4°C | |
Human IL-13 flex set (bead E6) |
Research use only