Learn More
BD Cytometric Bead Array (CBA) Human sCD154 (sCD40 Ligand); Flex Set, Bead C7
Bead-based immunoassay capable of measuring human soluble CD154, also known as sCD40 Ligand, in serum, plasma and cell culture supernatant samples
Brand:  BD 560305
Additional Details : Weight : 0.05000kg
Product Code. 15800099
  • 9895.00 SEK / Each
Estimated Shipment: 19-09-2024
to see stock.
Add to basket
Includes:
  • Human sCD154 (sCD40 Ligand) Standard
  • Human sCD154 (sCD40 Ligand) PE Detection Reagent
  • Human sCD154 (sCD40 Ligand) Capture Bead C7
Description
Description
Cotains one vial each of Capture Bead and PE Detection Reagent and two vials of Standard. The Capture Bead and PE Detection Reagent components of this flex set have been formulated to a 50x concentration to ensure product performance when multiplexed. The Standard component is lyophilized and should be transferred to a 15mL polypropylene tube for reconstitution. When reconstituted in 4.0mL Assay Diluent, the standard has a protein concentration of 2500pg/mL.
  • Human reactivity was determined by testing samples with CBA Human sCD154 (sCD40 Ligand) Flex Set
  • Must be used in conjunction with CBA Human Soluble Protein Master Buffer Kit, flow cytometer and FCAP Array Software
Specifications
Specifications
sCD154 (sCD40 Ligand)
Human sCD154 (sCD40 Ligand) standard, Human sCD154 (sCD40 Ligand) PE detection reagent, Human sCD154 (sCD40 Ligand) capture bead C7
4°C
Human sCD154 (sCD40 Ligand) flex set (bead C7)
Human
100 Tests
Flow Cytometry
Product Suggestions
Product Suggestions
Videos
Safety and Handling
Safety and Handling
Signal Word
  • Warning