Learn More
BD Cytometric Bead Array (CBA) Mouse IL-2; Flex Set, Bead A5
For detecting mouse interleukins in serum and culture samples
Brand: BD 558297
Additional Details : Weight : 0.10000kg
Includes: Mouse IL-2; Standard; Mouse IL-2; PE Detection Reagent; Mouse IL-2; Capture Bead A5
Description
The BD CBA Mouse IL-2 Flex Set is a bead-based immunoassay capable of measuring mouse interleukin-2 (IL-2) in serum and cell culture supernatant samples. Mouse reactivity was determined by testing samples with the BD CBA Mouse IL-2 Flex Set.
- Reactivity: Mouse
- Capture Bead and PE Detection Reagent components formulated to a 50x concentration for multiplex use
- Standard component is lyophilized
- When reconstituted in 4.0mL assay diluent, standard has a protein concentration of 2500pg/mL
- Must be used in conjunction with a BD CBA Human Soluble Protein Master Buffer Kit, flow cytometer, FCAP Array™ Software
Flow Cytometry
Specifications
IL-2 | |
Mouse IL-2, standard, Mouse IL-2, PE detection reagent, Mouse IL-2, capture bead A5 | |
Serum, Cell Culture Supernatant | |
Flow Cytometry |
Mouse | |
100 Tests | |
4°C | |
Mouse IL-2 flex set (bead A5) |
Safety and Handling
- Warning
Research Use Only