Learn More
BD D- Luciferin Sodium Salt
Specific for firefly luciferase in the illustrated reactions
Brand: BD 556876
Includes: Sodium salt
Description
Purified by HPLC techniques to remove dehydroluciferin and L-Luciferin, both of which are competitive inhibitors of firefly luciferase. Form dehydroluciferin in presence of ultraviolet light and/or moisture
- Synthetically made substrate
- lyophilized, in monosodium salt form
- Non-buffered solution
- Reconstitute lyophilized D-Luciferin with deionized water
Specifications
Lyophilized | |
D-Luciferin | |
-20°C |
Sodium salt | |
100 mg | |
Luciferase assay |