missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: B12 Assay Medium
For use in USP and AOAC methods for microbiological assay of Vitamin B12
Brand: BD 245710
This item is not returnable.
View return policy
Description
Specifications
ISO, 9000 | |
B12 Assay Medium | |
Vitamin assay Casamino Acids 15g/L, Dextrose 40g/L, Asparagine 0.2g/L, Sodium Acetate 20g/L, Ascorbic Acid 4g/L, L-Cystine 0.4g/L, DL-Tryptophan 0.4g/L, Adenine Sulfate 20mg/L, Guanine Hydrochloride 20mg/L, Uracil 20mg/L, Xanthine 20mg/L, Riboflavin 1mg/L, Thiamine Hydrochloride 1mg/L, Biotin 10μg/L, Niacin 2mg/L, p-Aminobenzoic Acid 2mg/L, Calcium Pantothenate 1mg/L, Pyridoxine Hydrochloride 4mg/L, Pyridoxal Hydrochloride 4mg/L, Pyridoxamine Hydrochloride 800μg/L, Folic Acid 200μg/L, Monopotassium Phosphate 1g/L, Dipotassium Phosphate 1g/L, Magnesium Sulfate 0.4g/L, Sodium Chloride 20mg/L, Ferrous Sulfate 20mg/L, Manganese Sulfate 20mg/L, Polysorbate 80 2g/L | |
Bottle |
Dehydrated | |
Microbiology Media | |
Isolation of Lactobacillus delbrueckii subspecies Lactis 7830 | |
100 g |
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction