missing translation for 'onlineSavingsMsg'
Learn More
Learn More
BD Difco™ Dehydrated Culture Media: Violet Red Bile Agar
For enumerating coliform organisms in dairy products
Brand: BD 211695
This item is not returnable.
View return policy
Description
- Formulation: Yeast extract 3g/L, Peptone 7g/L, No. 3 Bile Salt 1.5g/L, Lactose 10g/L, Sodium Chloride 5g/L, Agar 15g/L, Neutral red 0.03g/L, Crystal Violet 2mg/L
Order Info
Prices are for units of one each unless otherwise specified
Certifications
BAM, CCAM, COMPF, ISO, SMD
Specifications
BAM, CCAM, COMPF, ISO, SMD | |
2°C to 30°C | |
Violet Red Bile Agar | |
Yeast extract 3g/L, Peptone 7g/L, No. 3 Bile Salt 1.5g/L, Lactose 10g/L, Sodium Chloride 5g/L, Agar 15g/L, Neutral red 0.03g/L, Crystal Violet 2mg/L | |
Bottle |
Violet Red | |
Dehydrated | |
Agar | |
For enumerating coliform organisms in dairy products | |
500 g |
Safety and Handling
Signal Word
Hazard Category
Hazard Statement
Precautionary Statement
Supplemental information
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P272-Contaminated work clothing should not be allowed out of the workplace.
- P321-Specific treatment (see on this label).
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P302+P352-IF ON SKIN: Wash with plenty of water/.
- P362+P364-Take off contaminated clothing and wash it before reuse.
- P501-Dispose of contents/container to in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : oxgall
Product Content Correction
Your input is important to us. Please complete this form to provide feedback related to the content on this product.
Product Title
Spot an opportunity for improvement?Share a Content Correction