Learn More
BD ELISA Set II
Design an immunoassay with different sensitivity and dynamic range
Brand: BD 558534
Additional Details : Weight : 0.99000kg
Includes: Capture Antibody: Anti-mouse TNF-α, Detection Antibody: Biotinylated anti-mouse TNF-α, Enzyme Reagent: Streptavidin-horseradish peroxidase conjugate (SAv-HRP) (250X), Standard: Recombinant mouse TNF-α (lyophilized)
Description
- Flexible assay
- Calibrated against purified Baculovirus-expressed recombinant mouse TNF
ELISA
Specifications
15.6 to 1000pg/mL | |
20 96-well Plates | |
Biotin anti-Mouse TNF (250X) (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Mouse TNF (standard), Purified anti-Mouse TNF (250X) (capture) antibody, Streptavidin-HRP Conjugate (250X) | |
2°C to 8°C |
Mouse TNF ELISA set II | |
To develop ELISA for natural or recombinant Mouse TNF in Serum, Plasma and cell culture supernatants | |
20 Plates |
Safety and Handling
- Mouse TNF ELISA Set II
Signal Word
- Warning
Hazard Category
- Skin sensitiser Category 1
Hazard Statement
- H317-May cause an allergic skin reaction.
Precautionary Statement
- P261-Avoid breathing dust/fume/gas/mist/vapours/spray.
- P280-Wear protective gloves/protective clothing/eye protection/face protection.
- P333+P313-If skin irritation or rash occurs: Get medical advice/attention.
- P363-Wash contaminated clothing before reuse.
- P501b-Dispose of contents/container in accordance with local/regional/national/international regulations.
Supplemental information
- MIXTURE LIST-Contains : CMIT/MIT
Research use only.