Learn More
BD™ Falcon™ Roller Bottles, Easy On/Off Cap Product Code.: 11537411
BD™ Falcon™ Roller Bottles, Easy On/Off Cap
Product Code. 11537411
Polystyrene
Click to view available options
Material:
Polystyrene
Product Code. 11537411
Brand: BD Biosciences 353008
This item is currently unavailable or has been discontinued.
View the product page for possible alternatives.
View alternative products
Compatible with manual and automated filling systems
Ideal for use in research and manufacturing applications involving the scale-up of mammalian cells for purposes of virus propagation and bioproduct production.
  • One-piece BD Tufrol™ polystyrene manufactured using a proprietary process that provides high impact resistance
  • Sterilized by gamma irradiation, nonpyrogenic
Custom packaging and lot control available. Contact your Fisher Scientific Customer Service Representative.
TRUSTED_SUSTAINABILITY
Specifications
Bottle Surface Smooth
Material Polystyrene
Cell Growth Area 850 cm2
Product Content Correction
Product Title
BD™ Falcon™ Roller Bottles, Easy On/Off Cap >