Learn More
BD Falcon™ Roller Bottles, Vented Cap
Click to view available options
Pleated Surface; 1450cm2 Growth Area; Vented Cap; 20/cs double bag
Description
Ideal for use in research and manufacturing applications involving the scale-up of mammalian cells for purposes of virus propagation and bioproduct production.
- Available with smooth 850cm2 growth area configuration or pleated 1450cm2 growth area configuration
- One-piece BD Tufrol™ polystyrene manufactured using a proprietary process that provides high impact resistance
- Sterilized by gamma irradiation, nonpyrogenic
- Vented cap has integral hydrophobic 0.2μm membrane filter
Custom packaging and lot control available. Contact your Fisher Scientific Customer Service Representative.
Specifications
Bottle Surface | Pleated |
Material | Polystyrene |
Cell Growth Area | 1450 cm2 |