Learn More
BD Human GM-CSF ELISA Set
For human granulocyte macrophage colony stimulating factor (GM-CSF)
Brand: BD 555126
Additional Details : Weight : 0.01000kg
Description
The OptEIA™ set contains components necessary to develop enzyme-linked immunosorbent assays (ELISA) for natural or recombinant human GM-CSF in serum, plasma, and cell culture supernatants.
- Capture antibody: anti-human GM-CSF monoclonal antibody
- Detection antibody: biotinylated anti-human GM-CSF monoclonal antibody
- Enzyme reagent: Streptavidin-horseradish peroxidase conjugate
- Standards: lyophilized recombinant human GM-CSF
ELISA
Specifications
Human GM-CSF ELISA set | |
To develop ELISA for natural or recombinant Human GM-CSF in Serum, Plasma and cell culture supernatants | |
20 Plates |
20 96-well Plates | |
anti-Human GM-CSF (capture) Antibody, Biotinylated anti-Human GM-CSF (detection) Antibody, Instruction/analysis certificate, Lyophilized recombinant Human GM-CSF (standard), Streptavidin-HRP Conjugate | |
-80°C |
For research use only.