Learn More
BD Macro-Vue™ RPR Cards and Components
For large-scale testing in public health and clinical laboratories
2500.00 SEK
Specifications
For Use With (Application) | For large-scale testing in public health and clinical laboratories |
---|
Product Code | Brand | Quantity | Type | Price | Quantity & Availability | ||||
---|---|---|---|---|---|---|---|---|---|
Product Code | Brand | Quantity | Type | Price | Quantity & Availability | ||||
17352421
|
BD
276709 |
10 Pack | RPR Control Cards |
2500.00 SEK
Pack of 10 |
In Stock Log in to see stock. |
Please sign in to purchase this item. Need a web account? Register with us today! | |||
Description
RPR Card antigen suspension is a carbon particle cardiolipin antigen1 which detects reagin, an antibody-like substance present in serum or plasma from syphilitic persons, and occasionally in serum or plasma of persons with other acute or chronic conditions. The reagin binds to the test antigen, which consists of cardiolipin-lecithin-coated cholesterol particles, causing macroscopic flocculation.
- CLIA Complexity: Moderate
- Nontreponemal test
- For the serological detection of syphilis as described in A Manual of Tests for Syphilis†
Specifications
For large-scale testing in public health and clinical laboratories |