Learn More
BD Acridine Orange Reagent Dropper
Identify microorganisms without mixing or measuring reagents
Brand: BD 261182
Les retours ne sont pas autorisés pour ce produit.
Afficher la politique du retour.
Description
To perform tests, squeeze the dropper to crush the ampule. Invert the dropper for convenient drop-by-drop dispensing.
- Dropper contains one-day supply of reagent or stain, hermetically sealed inside an ampule
- Strict quality controls assure uniformity and accuracy
Spécification
Reagent/Stain Dropper | |
CE | |
Used for fluorescent microscopic detection of microorganisms in direct smears | |
Acridine Orange |
50 Pack | |
Dropper | |
15° to 30°C |