missing translation for 'onlineSavingsMsg'
Learn More

Behenic acid, tech. 85%, Thermo Scientific Chemicals

Product Code. 11495253
Click to view available options
Quantity:
250 g
1000 g
5000 g
This item is not returnable. View return policy
This item is not returnable. View return policy

Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.

This Thermo Scientific Chemicals brand product was originally part of the Alfa Aesar product portfolio. Some documentation and label information may refer to the legacy brand. The original Alfa Aesar product / item code or SKU reference has not changed as a part of the brand transition to Thermo Scientific Chemicals.

Applications
Behenic acid is used to give hair conditioners and moisturizers their smoothing properties. It is also used to investigate the phase behavior of long-chain acids in supercritical propane.

Solubility
Soluble in DMF (∼3 mg/ml), hot methanol, water (0.15 mg/ml at 25°C), chloroform, and ethanol (2.18 mg/ml at 25°C).

Notes
Store in cool place. Keep container tightly closed in a dry and well-ventilated place. Incompatible with strong oxidizing agents, reducing agents and bases. Store at room temperature.
TRUSTED_SUSTAINABILITY

Chemical Identifiers

CAS 112-85-6
Molecular Formula C22H44O2
Molecular Weight (g/mol) 340.592
MDL Number MFCD00002807
InChI Key UKMSUNONTOPOIO-UHFFFAOYSA-N
Synonym behenic acid, 1-docosanoic acid, n-docosanoic acid, hydrofol acid 560, docosoic acid, hydrofol 2022-55, glycon b-70, behensaeure, docosanoicacid, docosansaeure
PubChem CID 8215
ChEBI CHEBI:28941
IUPAC Name docosanoic acid
SMILES CCCCCCCCCCCCCCCCCCCCCC(=O)O

Specifications

Melting Point 74°C to 78°C
Boiling Point 306°C (60 mmHg)
Quantity 5000 g
Beilstein 1792887
Merck Index 14,1023
Solubility Information Soluble in DMF (∽3mg/ml),hot methanol,water (0.15mg/ml at 25°C),chloroform,and ethanol (2.18mg/ml at 25°C).
Formula Weight 340.6
Percent Purity 85%
Grade Technical
Chemical Name or Material Behenic acid

RUO – Research Use Only

missing translation for 'provideContentCorrection'

missing translation for 'productTitle'